5-(Trifluoromethyl)-o-anisidine ≥98.0%
Supplier: TCI America
Synonyms:
2-Methoxy-5-(trifluoromethyl)aniline, 5-(Trifluoromethyl)-2-anisidine, 5-(Trifluoromethyl)-2-methoxyaniline, 2-Amino-4-(trifluoromethyl)anisole, 3-Amino-4-methoxybenzotrifluoride
CAS Number: 349-65-5
MDL Number: MFCD00042486
Molecular Formula: C8H8F3NO
Molecular Weight: 191.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 60
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 3-Amino-4-methoxybenzotrifluoride, 2-Amino-4-(trifluoromethyl)anisole
Formula:
C₈H₈F₃NO MW: 191.15 g/mol Boiling Pt: 230 Melting Pt: 59 |
MDL Number:
MFCD00042486 CAS Number: 349-65-5 UN: 2811 ADR: 6.1 III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...