Methyl-4-hydroxybenzoate ≥99.0% (by HPLC, titration analysis) for biochemical research

Supplier: TCI America

Synonyms: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate

M2206-25G M2206-5G
TCM2206-25GEA 105.86 CAD
TCM2206-25G TCM2206-5G
Methyl-4-hydroxybenzoate ≥99.0% (by HPLC, titration analysis) for biochemical research
Methyl 4-hydroxybenzoate

[for Biochemical Research]
CAS Number: 99-76-3
MDL Number: MFCD00002352
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 280
Melting point (°C): 127
Flash Point (°C): 96


Product Categories: Bioscience, Preservatives for Biochemical Research
Other Product Names / Synonyms: 4-Hydroxybenzoic Acid Methyl Ester, Methylparaben

Formula: HOC₆H₄CO₂CH₃
Boiling Pt: 280 °C
Melting Pt: 127 °C
Flash Pt: 96 °C
MDL Number: MFCD00002352
CAS Number: 99-76-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f74611a5ada1021d8fb466ae6c95270e7a77bf2&vid=c62062663d2db318124fc169135c9b9198334e6b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR