Methyl-3-hydroxy-4-methoxybenzoate ≥98.0% (by GC)
Supplier: TCI America
M2205-25G
M2205-5G
TCM2205-25GEA
282.82
CAD
TCM2205-25G
TCM2205-5G
Methyl-3-hydroxy-4-methoxybenzoate ≥98.0% (by GC)
Methyl-3-hydroxy-4-methoxybenzoate
CAS Number: 6702-50-7
MDL Number: MFCD01321262
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 64
Other Product Names / Synonyms: Isovanillic Acid Methyl Ester, Methyl 3-Hydroxy-4-methoxybenzoate, 3-Hydroxy-4-methoxybenzoic Acid Methyl Ester
Formula:
C₉H₁₀O₄ Melting Pt: 64 °C |
MDL Number:
MFCD01321262 CAS Number: 6702-50-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...