Methyl-4-(aminomethyl)benzoate hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
4-(Aminomethyl)benzoic Acid Methyl Ester Hydrochloride, Methyl 4-(aminomethyl)benzoate hydrochloride
M2200-1G
M2200-5G
TCM2200-1GEA
53.72
CAD
TCM2200-1G
TCM2200-5G
Methyl-4-(aminomethyl)benzoate hydrochloride ≥98.0%
Methyl-4-(aminomethyl)benzoate hydrochloride
CAS Number: 6232-11-7
MDL Number: MFCD00182671
Molecular Formula: C9H11NO2
Molecular Weight: 201.65
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 238
Other Product Names / Synonyms: 4-(Aminomethyl)benzoic Acid Methyl Ester Hydrochloride
Formula:
C₉H₁₁NO₂ Melting Pt: 238 °C |
MDL Number:
MFCD00182671 CAS Number: 6232-11-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...