1-Methyl-4-[1-(4-piperidyl)-4-piperidyl]piperazine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M2197-25G
M2197-5G
TCM2197-25GEA
1297.18
CAD
TCM2197-25G
TCM2197-5G
1-Methyl-4-[1-(4-piperidyl)-4-piperidyl]piperazine ≥98.0% (by GC, titration analysis)
1-Methyl-4-[1-(4-piperidyl)-4-piperidyl]piperazine
CAS Number: 1629218-99-0
Molecular Formula: C15H30N4
Molecular Weight: 266.43
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 89
Other Product Names / Synonyms: 1-Methyl-4-(1,4'-bipiperidin-4-yl)piperazine
Formula:
C₁₅H₃₀N₄ Melting Pt: 89 °C |
MDL Number:
MFCD20257913 CAS Number: 1629218-99-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...