Methyl-3-(4-hydroxyphenyl)propionate ≥98.0%

Supplier: TCI America

Synonyms: 3-(4-Hydroxyphenyl)propionic Acid Methyl Ester, Methyl 3-(4-hydroxyphenyl)propanoate, Methyl 3-(4-hydroxyphenyl)propionate, 3-(4-Hydroxy-phenyl)-propionic acid-OMe

M2196-25G M2196-5G
TCM2196-25GEA 120.49 CAD
TCM2196-25G TCM2196-5G
Methyl-3-(4-hydroxyphenyl)propionate ≥98.0%
Methyl-3-(4-hydroxyphenyl)propionate

CAS Number: 5597-50-2
MDL Number: MFCD00071577
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 108
Melting point (°C): 40
Flash Point (°C): 113
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 3-(4-Hydroxyphenyl)propionic Acid Methyl Ester

Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
Boiling Pt: 108 °C
Melting Pt: 40 °C
Flash Pt: 113 °C
MDL Number: MFCD00071577
CAS Number: 5597-50-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77366ebbb7ec60c2cc091fdec07525a3636da265&vid=77366ebbb7ec60c2cc091fdec07525a3636da265&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR