Methyl-2-amino-3-methylbenzoate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 2-amino-3-methylbenzoate

M2193-25G M2193-5G
TCM2193-25GEA 262.28 CAD
TCM2193-25G TCM2193-5G
Methyl-2-amino-3-methylbenzoate ≥98.0%
Methyl 2-amino-3-methylbenzoate

CAS Number: 22223-49-0
MDL Number: MFCD06200918
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 153
Freezing point (°C): 27


Other Product Names / Synonyms: 2-Amino-3-methylbenzoic Acid Methyl Ester, Methyl 2-Amino-m-toluate, 2-Amino-m-toluic Acid Methyl Ester, Methyl 3-Methylanthranilate, 3-Methylanthranilic Acid Methyl Ester

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 153 °C
MDL Number: MFCD06200918
CAS Number: 22223-49-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5dde6d3765a77185de78e4cca96c4a281955b7c9&vid=5dde6d3765a77185de78e4cca96c4a281955b7c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR