6-Methoxypicolinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M2189-1G M2189-5G
TCM2189-1GEA 75.84 CAD
TCM2189-1G TCM2189-5G
6-Methoxypicolinic acid ≥98.0% (by GC, titration analysis)
6-Methoxypicolinic acid

CAS Number: 26893-73-2
MDL Number: MFCD06800962
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 134


Other Product Names / Synonyms: 6-Methoxypicolinic Acid

Formula: C₇H₇NO₃
Melting Pt: 134 °C
MDL Number: MFCD06800962
CAS Number: 26893-73-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9774680787ee5872ed6d9ad3f9e722cd19e46809&vid=9774680787ee5872ed6d9ad3f9e722cd19e46809&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR