4-(Methylamino)pyridine ≥98.0%

Supplier: TCI America

M2183-1G M2183-5G
TCM2183-1GEA 39.5 CAD
TCM2183-1G TCM2183-5G
4-(Methylamino)pyridine ≥98.0%
4-(Methylamino)pyridine

CAS Number: 1121-58-0
MDL Number: MFCD00152625
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 100
Melting point (°C): 127

Formula: C₆H₈N₂
Boiling Pt: 100 °C
Melting Pt: 127 °C
MDL Number: MFCD00152625
CAS Number: 1121-58-0
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94aeb7f0b55c2ef9d0b763ba1daef12413d73b3d&vid=94aeb7f0b55c2ef9d0b763ba1daef12413d73b3d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR