Methyl-3-bromo-4-methylbenzoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Methyl 3-bromo-4-methylbenzoate

M2181-25G M2181-5G
TCM2181-25GEA 176.8 CAD
TCM2181-25G TCM2181-5G
Methyl-3-bromo-4-methylbenzoate ≥98.0% (by GC)
Methyl 3-bromo-4-methylbenzoate

CAS Number: 104901-43-1
MDL Number: MFCD00144769
Molecular Formula: C9H9BrO2
Molecular Weight: 229.07
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 115
Specific Gravity (20/20): 1.47


Other Product Names / Synonyms: 3-Bromo-4-methylbenzoic Acid Methyl Ester, Methyl 3-Bromo-p-toluate, 3-Bromo-p-toluic Acid Methyl Ester

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 130 °C
Density: 1.47 (20 °C)
Flash Pt: 115 °C
MDL Number: MFCD00144769
CAS Number: 104901-43-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d78669bbb161f02efe5a8eb1954a188f530ef983&vid=d78669bbb161f02efe5a8eb1954a188f530ef983&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR