Methyl-4-benzyloxybenzoate ≥98.0%

Supplier: TCI America

M2180-25G M2180-5G
TCM2180-25GEA 836.45 CAD
TCM2180-25G TCM2180-5G
Methyl-4-benzyloxybenzoate ≥98.0%
Methyl 4-benzyloxybenzoate

CAS Number: 32122-11-5
MDL Number: MFCD00017613
Molecular Formula: C15H14O3
Molecular Weight: 242.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 99
Lambda max.: 257 nm (MeOH)


Other Product Names / Synonyms: 4-Benzyloxybenzoic Acid Methyl Ester

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Melting Pt: 99 °C
MDL Number: MFCD00017613
CAS Number: 32122-11-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e1256252024ed487fb180cd3587ae514a935a49&vid=7e1256252024ed487fb180cd3587ae514a935a49&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR