1-Methylbenzimidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M2173-25G M2173-5G
TCM2173-25GEA 369.72 CAD
TCM2173-25G TCM2173-5G
1-Methylbenzimidazole ≥98.0% (by GC, titration analysis)
1-Methylbenzimidazole

CAS Number: 1632-83-3
MDL Number: MFCD00192275
Molecular Formula: C8H8N2
Molecular Weight: 132.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 154
Melting point (°C): 59
Lambda max.: 277 nm (H2O)Storage Temperature: 0-10°C

Formula: C₈H₈N₂
Boiling Pt: 154 °C
Melting Pt: 59 °C
MDL Number: MFCD00192275
CAS Number: 1632-83-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1b86b75a6043d69690dd3dabf507a5f226d51cde&vid=1b86b75a6043d69690dd3dabf507a5f226d51cde&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR