Methyl-2-aminoisonicotinate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M2172-1G M2172-5G
TCM2172-1GEA 48.98 CAD
TCM2172-1G TCM2172-5G
Methyl-2-aminoisonicotinate ≥98.0% (by GC, titration analysis)
Methyl 2-aminoisonicotinate

CAS Number: 6937-03-7
MDL Number: MFCD04039316
Molecular Formula: C7H8N2O2
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 147


Other Product Names / Synonyms: 2-Aminoisonicotinic Acid Methyl Ester, Methyl 2-Aminopyridine-4-carboxylate, 2-Aminopyridine-4-carboxylic Acid Methyl Ester

Formula: C₇H₈N₂O₂
Melting Pt: 147 °C
MDL Number: MFCD04039316
CAS Number: 6937-03-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18a19e54d5723d1a4c3298dcc695bd47091306da&vid=18a19e54d5723d1a4c3298dcc695bd47091306da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR