1-(2-Methoxyethyl)piperazine ≥98.0%

Supplier: TCI America

M2169-1G M2169-5G
TCM2169-1GEA 129.56 CAD
TCM2169-1G TCM2169-5G
1-(2-Methoxyethyl)piperazine ≥98.0%
1-(2-Methoxyethyl)piperazine

CAS Number: 13484-40-7
MDL Number: MFCD00191214
Molecular Formula: C7H16N2O
Molecular Weight: 144.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 101
Specific Gravity (20/20): 0.97


Other Product Names / Synonyms: Methyl 2-(1-Piperazinyl)ethyl Ether

Formula: C₇H₁₆N₂O
Boiling Pt: 101 °C
Density: 0.97 (20 °C)
MDL Number: MFCD00191214
CAS Number: 13484-40-7
UN: 3267
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7433556d13df424ebdc470c86ddcf4f4ca8e3ac&vid=e7433556d13df424ebdc470c86ddcf4f4ca8e3ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR