3-Mercapto-1-hexanol ≥98.0%

Supplier: TCI America

Synonyms: 3-Mercaptohexan-1-ol

M2168-25G M2168-5G
TCM2168-25GEA 347.13 CAD
TCM2168-25G TCM2168-5G
3-Mercapto-1-hexanol ≥98.0%
3-Mercapto-1-hexanol

CAS Number: 51755-83-0
MDL Number: MFCD00792515
Molecular Formula: C6H14OS
Molecular Weight: 134.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 45
Flash Point (°C): 100
Specific Gravity (20/20): 0.98

Formula: C₆H₁₄OS
MW: 134.24 g/mol
Boiling Pt: 45
Density: 0.98 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00792515
CAS Number: 51755-83-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3400656ba98cfbd3c2fb038a5583eae2c8a0b5b5&vid=3400656ba98cfbd3c2fb038a5583eae2c8a0b5b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR