Methyl-3-methoxysalicylate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 2-hydroxy-3-methoxybenzoate

M2167-25G M2167-5G
TCM2167-25GEA 419.81 CAD
TCM2167-25G TCM2167-5G
Methyl-3-methoxysalicylate ≥98.0%
Methyl 3-methoxysalicylate

CAS Number: 6342-70-7
MDL Number: MFCD00239511
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 68
Lambda max.: 320 nm (Dioxane)


Other Product Names / Synonyms: 3-Methoxysalicylic Acid Methyl Ester, Methyl 2-Hydroxy-3-methoxybenzoate, 2-Hydroxy-3-methoxybenzoic Acid Methyl Ester, Methyl 2-Hydroxy-m-anisate, 2-Hydroxy-m-anisic Acid Methyl Ester

Formula: C₉H₁₀O₄
Melting Pt: 68 °C
MDL Number: MFCD00239511
CAS Number: 6342-70-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aab69d91af148335f4d7eae803d3689c59c7b5aa&vid=aab69d91af148335f4d7eae803d3689c59c7b5aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR