Methyl-2-chloronicotinate ≥98.0% (by GC)
Supplier: TCI America
M2165-25G
M2165-5G
TCM2165-25GEA
79
CAD
TCM2165-25G
TCM2165-5G
Methyl-2-chloronicotinate ≥98.0% (by GC)
Methyl 2-chloronicotinate
CAS Number: 40134-18-7
MDL Number: MFCD01318629
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 70
Flash Point (°C): 110
Specific Gravity (20/20): 1.32
Other Product Names / Synonyms: 2-Chloronicotinic Acid Methyl Ester, Methyl 2-Chloro-3-pyridinecarboxylate, 2-Chloro-3-pyridinecarboxylic Acid Methyl Ester
Formula:
C₇H₆ClNO₂ Boiling Pt: 70 °C Density: 1.32 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD01318629 CAS Number: 40134-18-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...