Methyl-2-chloroisonicotinate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 2-chloropyridine-4-carboxylate, Methyl 2-chloroisonicotinate

M2164-25G M2164-5G
TCM2164-25GEA 110.6 CAD
TCM2164-25G TCM2164-5G
Methyl-2-chloroisonicotinate ≥98.0%
Methyl 2-chloroisonicotinate

CAS Number: 58481-11-1
MDL Number: MFCD01765409
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 70
Melting point (°C): 34
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Chloroisonicotinic Acid Methyl Ester, Methyl 2-Chloro-4-pyridinecarboxylate, 2-Chloro-4-pyridinecarboxylic Acid Methyl Ester

Formula: C₇H₆ClNO₂
MW: 171.5810 g/mol
Boiling Pt: 70 °C
Melting Pt: 34 °C
MDL Number: MFCD01765409
CAS Number: 58481-11-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b6b13c8ac902301fb0ad0d85f8ec0cbc53d4411&vid=0b6b13c8ac902301fb0ad0d85f8ec0cbc53d4411&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR