Methyl-4-acetylbenzoate ≥98.0% (by GC)
Supplier: TCI America
M2163-25G
M2163-5G
TCM2163-25GEA
369.72
CAD
TCM2163-25G
TCM2163-5G
Methyl-4-acetylbenzoate ≥98.0% (by GC)
Methyl 4-acetylbenzoate
CAS Number: 3609-53-8
MDL Number: MFCD00216474
Molecular Formula: C10H10O3
Molecular Weight: 178.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 97
Other Product Names / Synonyms: 4-Acetylbenzoic Acid Methyl Ester
Formula:
C₁₀H₁₀O₃ Melting Pt: 97 °C |
MDL Number:
MFCD00216474 CAS Number: 3609-53-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...