4-Methoxy-2-(trifluoromethyl)benzaldehyde ≥97.0%

Supplier: TCI America

M2162-1G M2162-5G
TCM2162-1GEA 91.64 CAD
TCM2162-1G TCM2162-5G
4-Methoxy-2-(trifluoromethyl)benzaldehyde ≥97.0%
4-Methoxy-2-(trifluoromethyl)benzaldehyde

CAS Number: 106312-36-1
MDL Number: MFCD01091011
Molecular Formula: C9H7F3O2
Molecular Weight: 204.15
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 40
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Formyl-3-(trifluoromethyl)anisole

Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Melting Pt: 40 °C
MDL Number: MFCD01091011
CAS Number: 106312-36-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc96bcdeecec688dd3fda01474cb0b4e1e089874&vid=cc96bcdeecec688dd3fda01474cb0b4e1e089874&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR