Methyl-1H-pyrrole-2-carboxylate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Methyl Pyrrole-2-carboxylate, Methyl 1H-pyrrole-2-carboxylate, Pyrrole-2-carboxylic acid methyll ester, Methyl pyrrole-2-carboxylate
M2160-25G
M2160-5G
TCM2160-25GEA
233.84
CAD
TCM2160-25G
TCM2160-5G
Methyl-1H-pyrrole-2-carboxylate ≥98.0% (by GC)
Methyl-1H-pyrrole-2-carboxylate
CAS Number: 1193-62-0
MDL Number: MFCD00817048
Molecular Formula: C6H7NO2
Molecular Weight: 125.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 73
Other Product Names / Synonyms: Pyrrole-2-carboxylic Acid Methyl Ester
Formula:
C₆H₇NO₂ MW: 125.13 g/mol Melting Pt: 73 °C |
MDL Number:
MFCD00817048 CAS Number: 1193-62-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...