2-Methoxy-3-pyridineboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 2-Methoxypyridine-3-boronic acid

M2159-1G M2159-5G
TCM2159-1GEA 54.67 CAD
TCM2159-1G TCM2159-5G
2-Methoxy-3-pyridineboronic acid (contains varying amounts of Anhydride)
2-Methoxy-3-pyridineboronic acid

CAS Number: 163105-90-6
MDL Number: MFCD03411572
Molecular Formula: C6H8BNO3
Molecular Weight: 152.94
Form: Crystal
Color: White
Melting point (°C): 145
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Methoxy-3-pyridylboronic Acid

Formula: C₆H₈BNO₃
MW: 152.95 g/mol
Melting Pt: 145 °C
MDL Number: MFCD03411572
CAS Number: 163105-90-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=982d96e4ca338ed34b58f2b3952f1a50e6590efc&vid=982d96e4ca338ed34b58f2b3952f1a50e6590efc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR