(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M2157-25G M2157-5G
TCM2157-25GEA 266.07 CAD
TCM2157-25G TCM2157-5G
(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol ≥98.0% (by GC, titration analysis)
(S)-(-)-3-(N-Methylamino)-1-(2-thienyl)-1-propanol

CAS Number: 116539-55-0
MDL Number: MFCD07357308
Molecular Formula: C8H13NOS
Molecular Weight: 171.26
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 74
Specific rotation [a]20/D: -12 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-2-[3-(Methylamino)-1-hydroxypropyl]thiophene

Formula: C₈H₁₃NOS
Melting Pt: 74 °C
MDL Number: MFCD07357308
CAS Number: 116539-55-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=633c44d31a946765c7fe93ceaf21768b32ce9f6d&vid=633c44d31a946765c7fe93ceaf21768b32ce9f6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR