1-Methyl-1H-imidazole-4,5-dicarboxylic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI America
M2156-1G
M2156-5G
TCM2156-1GEA
154.84
CAD
TCM2156-1G
TCM2156-5G
1-Methyl-1H-imidazole-4,5-dicarboxylic acid ≥97.0% (by GC, titration analysis)
1-Methyl-1H-imidazole-4,5-dicarboxylic acid
CAS Number: 19485-38-2
MDL Number: MFCD01552045
Molecular Formula: C6H6N2O4
Molecular Weight: 170.12
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 262
Other Product Names / Synonyms: 4,5-Dicarboxy-1-methyl-1H-imidazole
Formula:
C₆H₆N₂O₄ Melting Pt: 262 °C |
MDL Number:
MFCD01552045 CAS Number: 19485-38-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...