Methyl-4-amino-3-methylbenzoate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 4-amino-3-methylbenzoate

M2152-25G M2152-5G
TCM2152-25GEA 265.44 CAD
TCM2152-25G TCM2152-5G
Methyl-4-amino-3-methylbenzoate ≥98.0%
Methyl 4-amino-3-methylbenzoate

CAS Number: 18595-14-7
MDL Number: MFCD00102230
Molecular Formula: C9H11NO2
Molecular Weight: 165.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 120


Other Product Names / Synonyms: 4-Amino-3-methylbenzoic Acid Methyl Ester, Methyl 4-Amino-m-toluate, 4-Amino-m-toluic Acid Methyl Ester

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Melting Pt: 120 °C
MDL Number: MFCD00102230
CAS Number: 18595-14-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=260edfe26687462247ddc0dbeca7d618fa02cb44&vid=260edfe26687462247ddc0dbeca7d618fa02cb44&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR