Potassium methyl malonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Potassium 3-methoxy-3-oxopropanoate, Monomethyl monopotassium malonate, Monomethyl malonate potassium salt

M2149-250G M2149-25G
TCM2149-250GEA 382.36 CAD
TCM2149-250G TCM2149-25G
Potassium methyl malonate ≥98.0% (by titrimetric analysis)
Potassium methyl malonate

CAS Number: 38330-80-2
MDL Number: MFCD00014021
Molecular Formula: C4H6O4
Molecular Weight: 156.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 162


Other Product Names / Synonyms: Methyl Potassium Malonate, Malonic Acid Methyl Ester Potassium Salt, Malonic Acid Monomethyl Ester Potassium Salt, Monomethyl Malonate Potassium Salt

Formula: C₄H₅KO₄
MW: 156.18 g/mol
Melting Pt: 162 °C
MDL Number: MFCD00014021
CAS Number: 38330-80-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c247d54f0b9c618a177bdc51507c3b8368ed3669&vid=c247d54f0b9c618a177bdc51507c3b8368ed3669&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR