Methyl-4-methyl-3-nitrobenzoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Methyl 4-methyl-3-nitrobenzoate

M2146-25G M2146-5G
TCM2146-25GEA 171.9 CAD
TCM2146-25G TCM2146-5G
Methyl-4-methyl-3-nitrobenzoate ≥98.0% (by GC)
Methyl 4-methyl-3-nitrobenzoate

CAS Number: 7356-11-8
MDL Number: MFCD00130054
Molecular Formula: C9H9NO4
Molecular Weight: 195.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 51
Flash Point (°C): 110


Other Product Names / Synonyms: 4-Methyl-3-nitrobenzoic Acid Methyl Ester, Methyl 3-Nitro-p-toluate, 3-Nitro-p-toluic Acid Methyl Ester

Formula: C₉H₉NO₄
MW: 195.18 g/mol
Melting Pt: 51 °C
Flash Pt: 110 °C
MDL Number: MFCD00130054
CAS Number: 7356-11-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b21a08fbae5b1ef08033c43f36def2faa01df2ad&vid=b21a08fbae5b1ef08033c43f36def2faa01df2ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR