Methyl-3-methyl-4-nitrobenzoate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 3-methyl-4-nitrobenzoate

M2145-25G M2145-5G
TCM2145-25GEA 223.1 CAD
TCM2145-25G TCM2145-5G
Methyl-3-methyl-4-nitrobenzoate ≥98.0%
Methyl 3-methyl-4-nitrobenzoate

CAS Number: 24078-21-5
MDL Number: MFCD00085640
Molecular Formula: C9H9NO4
Molecular Weight: 195.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 81


Other Product Names / Synonyms: 3-Methyl-4-nitrobenzoic Acid Methyl Ester, Methyl 4-Nitro-m-toluate, 4-Nitro-m-toluic Acid Methyl Ester

Formula: C₉H₉NO₄
MW: 195.18 g/mol
Melting Pt: 81 °C
MDL Number: MFCD00085640
CAS Number: 24078-21-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=859903624bc55cf8b46e0e58a5e19725a228a2af&vid=859903624bc55cf8b46e0e58a5e19725a228a2af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR