Methyl-2-methyl-3-nitrobenzoate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 2-methyl-3-nitrobenzoate

M2144-25G M2144-5G
TCM2144-25GEA 67.94 CAD
TCM2144-25G TCM2144-5G
Methyl-2-methyl-3-nitrobenzoate ≥98.0%
Methyl 2-methyl-3-nitrobenzoate

CAS Number: 59382-59-1
MDL Number: MFCD00082564
Molecular Formula: C9H9NO4
Molecular Weight: 195.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 67


Other Product Names / Synonyms: 2-Methyl-3-nitrobenzoic Acid Methyl Ester, Methyl 3-Nitro-o-toluate, 3-Nitro-o-toluic Acid Methyl Ester

Formula: C₉H₉NO₄
MW: 195.17 g/mol
Melting Pt: 67 °C
MDL Number: MFCD00082564
CAS Number: 59382-59-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=22b98218b11719b1bfefb6272cfea792e1c72136&vid=22b98218b11719b1bfefb6272cfea792e1c72136&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR