(3-Chlorophenyl)acetic acid methyl ester ≥98.0% (by GC)

Supplier: TCI America

M2142-25G M2142-5G
TCM2142-25GEA 227.52 CAD
TCM2142-25G TCM2142-5G
(3-Chlorophenyl)acetic acid methyl ester ≥98.0% (by GC)
(3-Chlorophenyl)acetic acid methyl ester

CAS Number: 53088-68-9
MDL Number: MFCD00800679
Molecular Formula: C9H9ClO2
Molecular Weight: 184.62
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.20


Other Product Names / Synonyms: (3-Chlorophenyl)acetic Acid Methyl Ester

Formula: (C₆H₄Cl)CH₂COOCH₃
Density: 1.20 (20 °C)
MDL Number: MFCD00800679
CAS Number: 53088-68-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1061deb0cfbf54bee48303ee67049c057f3ba2f2&vid=1061deb0cfbf54bee48303ee67049c057f3ba2f2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR