2-(4-Methoxystyryl)-4,6-bis(trichloromethyl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

M2140-5G M2140-25G
TCM2140-5GEA 162.39 CAD
TCM2140-5G TCM2140-25G
2-(4-Methoxystyryl)-4,6-bis(trichloromethyl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2-(4-Methoxystyryl)-4,6-bis(trichloromethyl)-1,3,5-triazine

CAS Number: 42573-57-9
MDL Number: MFCD01940892
Molecular Formula: C14H9Cl6N3O
Molecular Weight: 447.95
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 193

Formula: C₁₄H₉Cl₆N₃O
MW: 447.95 g/mol
Melting Pt: 193 °C
MDL Number: MFCD01940892
CAS Number: 42573-57-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfe649fe746bb103ec65009634497e00a6992294&vid=dfe649fe746bb103ec65009634497e00a6992294&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR