3,5-Dimethyl-p-anisic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3,5-Dimethyl-p-anisic acid, 4-Methoxy-3,5-dimethylbenzoic acid, 3,5-Dimethyl-4-methoxybenzenecarboxylic acid, 3,5-Dimethyl-4-methoxybenzoic acid

M2139-1G M2139-5G
TCM2139-1GEA 64.78 CAD
TCM2139-1G TCM2139-5G
3,5-Dimethyl-p-anisic acid ≥98.0% (by GC, titration analysis)
3,5-Dimethyl-p-anisic acid

CAS Number: 21553-46-8
MDL Number: MFCD00020309
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 194


Other Product Names / Synonyms: 3,5-Dimethyl-p-anisic Acid

Formula: C₁₀H₁₂O₃
MW: 180.2
Boiling Pt: 313
Melting Pt: 194 °C
Density: 1.253
MDL Number: MFCD00020309
CAS Number: 21553-46-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b635d63d86cf8c94cf113c65aab7e2bd0ae35994&vid=b635d63d86cf8c94cf113c65aab7e2bd0ae35994&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR