Methyl-3-chloro-4-methylbenzoate ≥98.0% (by GC)
Supplier: TCI America
M2136-25G
M2136-5G
TCM2136-25GEA
235.42
CAD
TCM2136-25G
TCM2136-5G
Methyl-3-chloro-4-methylbenzoate ≥98.0% (by GC)
Methyl 3-chloro-4-methylbenzoate
CAS Number: 56525-63-4
MDL Number: MFCD00205141
Molecular Formula: C9H9ClO2
Molecular Weight: 184.62
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 107
Freezing point (°C): 28
Other Product Names / Synonyms: 3-Chloro-4-methylbenzoic Acid Methyl Ester, Methyl 3-Chloro-p-toluate, 3-Chloro-p-toluic Acid Methyl Ester
Formula:
C₉H₉ClO₂ Boiling Pt: 107 °C |
MDL Number:
MFCD00205141 CAS Number: 56525-63-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...