Methyl-5-methoxysalicylate ≥98.0%

Supplier: TCI America

Synonyms: Methyl 2-hydroxy-5-methoxybenzoate

M2127-25G M2127-5G
TCM2127-25GEA 243.32 CAD
TCM2127-25G TCM2127-5G
Methyl-5-methoxysalicylate ≥98.0%
Methyl 5-methoxysalicylate

CAS Number: 2905-82-0
MDL Number: MFCD00017185
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 240
Flash Point (°C): 113
Specific Gravity (20/20): 1.23
Lambda max.: 334 nm (CH3CN)


Other Product Names / Synonyms: 5-Methoxysalicylic Acid Methyl Ester, Methyl 2-Hydroxy-5-methoxybenzoate, 2-Hydroxy-5-methoxybenzoic Acid Methyl Ester

Formula: C₉H₁₀O₄
Boiling Pt: 240 °C
Density: 1.23 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00017185
CAS Number: 2905-82-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69ba0da466d1311333f97619aa2c2cbbad630c7b&vid=69ba0da466d1311333f97619aa2c2cbbad630c7b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR