Methyl-2-aminonicotinate ≥97.0% (by GC, titration analysis)
Supplier: TCI America
M2125-1G
M2125-5G
TCM2125-1GEA
47.4
CAD
TCM2125-1G
TCM2125-5G
Methyl-2-aminonicotinate ≥97.0% (by GC, titration analysis)
Methyl 2-aminonicotinate
CAS Number: 14667-47-1
MDL Number: MFCD00128872
Molecular Formula: C7H8N2O2
Molecular Weight: 152.15
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 84
Other Product Names / Synonyms: 2-Aminonicotinic Acid Methyl Ester, Methyl 2-Aminopyridine-3-carboxylate, 2-Aminopyridine-3-carboxylic Acid Methyl Ester
Formula:
C₇H₈N₂O₂ Melting Pt: 84 °C |
MDL Number:
MFCD00128872 CAS Number: 14667-47-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...