Methyl-2-bromo-4-nitrobenzoate ≥98.0% (by GC)

Supplier: TCI America

M2123-1G M2123-5G
TCM2123-1GEA 31.6 CAD
TCM2123-1G TCM2123-5G
Methyl-2-bromo-4-nitrobenzoate ≥98.0% (by GC)
Methyl 2-bromo-4-nitrobenzoate

CAS Number: 100959-22-6
MDL Number: MFCD07779271
Molecular Formula: C8H6BrNO4
Molecular Weight: 260.04
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 84


Other Product Names / Synonyms: 2-Bromo-4-nitrobenzoic Acid Methyl Ester

Formula: C₈H₆BrNO₄
Melting Pt: 84 °C
MDL Number: MFCD07779271
CAS Number: 100959-22-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9ff710455a53303e3d0ca5efe0c4b56587c4b26&vid=f9ff710455a53303e3d0ca5efe0c4b56587c4b26&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR