Matrine ≥98.0%

Supplier: TCI America

Synonyms: (7aS,13aR,13bR,13cS)-Dodecahydro-1H,5H,10H-dipyrido[2,1-f:3',2',1'-ij][1,6]naphthyridin-10-one, Matridin-15-one

M2120-1G M2120-200MG
TCM2120-1GEA 914.03 CAD
TCM2120-1G TCM2120-200MG
Matrine ≥98.0%
Matrine

CAS Number: 519-02-8
MDL Number: MFCD00210527
Molecular Formula: C15H24N2O
Molecular Weight: 248.37
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 223
Melting point (°C): 85
Specific rotation [a]20/D: 33 deg (C=1, EtOH)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Alkaloids (Phytochemicals), Piperidine Alkaloids
Other Product Names / Synonyms: Matridin-15-one, (7aS,13aR,13bR,13cS)-Dodecahydro-1H,5H,10H-dipyrido[2,1-f:3',2',1'-ij][1,6]naphthyridin-10-one

Formula: C₁₅H₂₄N₂O
MW: 248.37 g/mol
Boiling Pt: 223 °C
Melting Pt: 85 °C
MDL Number: MFCD00210527
CAS Number: 519-02-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=128fcc6b6adbeb9d3fa928de04fe5ff7ac61c34c&vid=128fcc6b6adbeb9d3fa928de04fe5ff7ac61c34c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR