Methyl-1-adamantanecarboxylate ≥98.0% (by GC)

Supplier: TCI America

M2119-25G M2119-5G
TCM2119-25GEA 549.84 CAD
TCM2119-25G TCM2119-5G
Methyl-1-adamantanecarboxylate ≥98.0% (by GC)
Methyl-1-adamantanecarboxylate
Formula: C₁₂H₁₈O₂
Boiling Pt: 79 °C
Melting Pt: 37 °C
MDL Number: MFCD01838519
CAS Number: 711-01-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b98509eae739e16554d7ba0d0117004d707d292d&vid=b98509eae739e16554d7ba0d0117004d707d292d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR