4-[(4-Methyl-1-piperazinyl)methyl]benzoic acid dihydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 4-[(4-Methylpiperazin-1-yl)methyl]benzoic acid dihydrochloride

M2110-25G M2110-5G
TCM2110-25GEA 551.42 CAD
TCM2110-25G TCM2110-5G
4-[(4-Methyl-1-piperazinyl)methyl]benzoic acid dihydrochloride ≥98.0%
4-[(4-Methyl-1-piperazinyl)methyl]benzoic acid dihydrochloride

CAS Number: 106261-49-8
MDL Number: MFCD07772867
Molecular Formula: C13H18N2O2
Molecular Weight: 307.22
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal

Formula: C₁₃H₁₈N₂O₂
MW: 307.22 g/mol
MDL Number: MFCD07772867
CAS Number: 106261-49-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b160c156c3801ec4edca6dd766de2709bda16679&vid=b160c156c3801ec4edca6dd766de2709bda16679&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR