Methyl-6-aminonicotinate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M2107-25G
M2107-5G
TCM2107-25GEA
447.93
CAD
TCM2107-25G
TCM2107-5G
Methyl-6-aminonicotinate ≥98.0% (by GC, titration analysis)
Methyl 6-aminonicotinate
CAS Number: 36052-24-1
MDL Number: MFCD00797844
Molecular Formula: C7H8N2O2
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 160
Other Product Names / Synonyms: 6-Aminonicotinic Acid Methyl Ester, Methyl 6-Aminopyridine-3-carboxylate, 6-Aminopyridine-3-carboxylic Acid Methyl Ester
Formula:
C₇H₈N₂O₂ Melting Pt: 160 °C |
MDL Number:
MFCD00797844 CAS Number: 36052-24-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...