4-Methoxyphenyl-4-(3-butenyloxy)benzoate ≥98.0% (by GC)

Supplier: TCI America

M2106-1G M2106-5G
TCM2106-1GEA 93.22 CAD
TCM2106-1G TCM2106-5G
4-Methoxyphenyl-4-(3-butenyloxy)benzoate ≥98.0% (by GC)
4-Methoxyphenyl-4-(3-butenyloxy)benzoate

CAS Number: 76487-56-4
MDL Number: MFCD00143337
Molecular Formula: C18H18O4
Molecular Weight: 298.34
Purity/Analysis Method: >98.0% (GC)
Form: Crystal


Product Categories: Materials Chemistry, Liquid Crystal Materials (Optically Functional Materials)
Other Product Names / Synonyms: 4-(3-Butenyloxy)benzoic Acid 4-Methoxyphenyl Ester

Formula: C₁₈H₁₈O₄
MDL Number: MFCD00143337
CAS Number: 76487-56-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0227c442f700bb721b6a8fc37d508cd8e46260c5&vid=0227c442f700bb721b6a8fc37d508cd8e46260c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR