2-(4-Methoxyphenyl)pyridine ≥97.0%

Supplier: TCI America

Synonyms: 4-(2-Pyridyl)anisole

M2101-1G M2101-25G M2101-5G
TCM2101-1GEA 67.94 CAD
TCM2101-1G TCM2101-25G TCM2101-5G
2-(4-Methoxyphenyl)pyridine ≥97.0%
2-(4-Methoxyphenyl)pyridine

CAS Number: 5957-90-4
MDL Number: MFCD00092333
Molecular Formula: C12H11NO
Molecular Weight: 185.23
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Boiling point (°C): 130
Melting point (°C): 55
Lambda max.: 282 nm (MeOH)


Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 4-(2-Pyridyl)anisole

Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
Boiling Pt: 130 °C
Melting Pt: 55 °C
MDL Number: MFCD00092333
CAS Number: 5957-90-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e09a3254c929d2eed290151b00f00e280ad677b&vid=7e09a3254c929d2eed290151b00f00e280ad677b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR