7-Methoxy-1-tetralone ≥98.0%
Supplier: TCI America
Synonyms:
7-Methoxy-3,4-dihydronaphthalen-1(2H)-one
M2092-25G
M2092-5G
TCM2092-25GEA
261.98
CAD
TCM2092-25G
TCM2092-5G
7-Methoxy-1-tetralone ≥98.0%
7-Methoxy-1-tetralone
CAS Number: 6836-19-7
MDL Number: MFCD00001696
Molecular Formula: C11H12O2
Molecular Weight: 176.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 62
Other Product Names / Synonyms: 3,4-Dihydro-7-methoxy-1(2H)-naphthalenone
Formula:
C₁₁H₁₂O₂ MW: 176.22 g/mol Melting Pt: 62 °C |
MDL Number:
MFCD00001696 CAS Number: 6836-19-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...