4-Methoxyphenyl-2-O-acetyl-3-O-allyl-4,6-O-benzylidene-β-D-glucopyranoside ≥98.0%
Supplier: TCI America
M2065-1G
M2065-5G
TCM2065-1GEA
309.05
CAD
TCM2065-1G
TCM2065-5G
4-Methoxyphenyl-2-O-acetyl-3-O-allyl-4,6-O-benzylidene-β-D-glucopyranoside ≥98.0%
4-Methoxyphenyl-2-O-acetyl-3-O-allyl-4,6-O-benzylidene-beta-D-glucopyranoside
CAS Number: 1477956-18-5
Molecular Formula: C25H28O8
Molecular Weight: 456.49
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: -23 deg (C=1, CHCl3)
Storage Temperature: 0-10°C
Formula:
C₂₅H₂₈O₈ |
CAS Number:
1477956-18-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...