2-Methyltryptamine ≥98.0%

Supplier: TCI America

Synonyms: 3-(2-Aminoethyl)-2-methylindole

M2055-1G M2055-5G
TCM2055-1GEA 251.22 CAD
TCM2055-1G TCM2055-5G
2-Methyltryptamine ≥98.0%
2-Methyltryptamine

CAS Number: 2731-06-8
MDL Number: MFCD00130185
Molecular Formula: C11H14N2
Molecular Weight: 174.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 177
Melting point (°C): 108
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 3-(2-Aminoethyl)-2-methylindole

Formula: C₁₁H₁₄N₂
Boiling Pt: 177 °C
Melting Pt: 108 °C
MDL Number: MFCD00130185
CAS Number: 2731-06-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a64d64f32f39d3038bb9990bedf11137a88a39e&vid=5a64d64f32f39d3038bb9990bedf11137a88a39e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR