5-Methylpyrazine-2-carboxylic acid-4-oxide ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Acipimox, 5-Carboxy-2-methylpyrazine 1-oxide, 2-Carboxy-5-methylpyrazine 4-Oxide, 5-Carboxy-2-methylpyrazine 1-Oxide

M2053-1G M2053-5G
TCM2053-1GEA 110.76 CAD
TCM2053-1G TCM2053-5G
5-Methylpyrazine-2-carboxylic acid-4-oxide ≥98.0% (by GC)
5-Methylpyrazine-2-carboxylic acid-4-oxide

CAS Number: 51037-30-0
MDL Number: MFCD00865757
Molecular Formula: C6H6N2O3
Molecular Weight: 154.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 180
Storage Temperature: 0-10°C


Product Categories: Bioscience, Antilipemic Ingredients
Other Product Names / Synonyms: 2-Carboxy-5-methylpyrazine 4-Oxide, 5-Carboxy-2-methylpyrazine 1-Oxide, Acipimox

Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 180 °C
MDL Number: MFCD00865757
CAS Number: 51037-30-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a7e72b26667efcd405f6bbcf1f45ccf8f6974be&vid=8a7e72b26667efcd405f6bbcf1f45ccf8f6974be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR