Methyl trans-4-hydroxycyclohexanecarboxylate ≥98.0% (by GC)
Supplier: TCI America
M1991-5G
M1991-1G
TCM1991-5GEA
339.7
CAD
TCM1991-5G
TCM1991-1G
Methyl trans-4-hydroxycyclohexanecarboxylate ≥98.0% (by GC)
Methyl trans-4-hydroxycyclohexanecarboxylate
CAS Number: 6125-57-1
Molecular Formula: C8H14O3
Molecular Weight: 158.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 125
Specific Gravity (20/20): 1.10
Other Product Names / Synonyms: trans-4-Hydroxycyclohexanecarboxylic Acid Methyl Ester
Formula:
C₈H₁₄O₃ Boiling Pt: 125 °C Density: 1.10 (20 °C) |
MDL Number:
MFCD20441963 CAS Number: 6125-57-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...