Methyl-4-(aminomethyl)cyclohexanecarboxylate hydrochloride (cis and trans mixture) ≥98.0% (by titrimetric analysis)
Supplier: TCI America
M1987-1G
M1987-5G
TCM1987-1GEA
203.35
CAD
TCM1987-1G
TCM1987-5G
Methyl-4-(aminomethyl)cyclohexanecarboxylate hydrochloride (cis and trans mixture) ≥98.0% (by titrimetric analysis)
Methyl-4-(aminomethyl)cyclohexanecarboxylate hydrochloride (cis and trans mixture)
CAS Number: 54640-02-7
MDL Number: MFCD03093748
Molecular Formula: C9H17NO2
Molecular Weight: 207.70
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: 4-(Aminomethyl)cyclohexanecarboxylic Acid Methyl Ester Hydrochloride
Formula:
C₉H₁₇NO₂ |
MDL Number:
MFCD03093748 CAS Number: 54640-02-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...