1-Methyl-1-propylpiperidinium bromide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
M1976-25G
M1976-5G
TCM1976-25GEA
448.72
CAD
TCM1976-25G
TCM1976-5G
1-Methyl-1-propylpiperidinium bromide ≥98.0% (by titrimetric analysis)
1-Methyl-1-propylpiperidinium bromide
CAS Number: 88840-42-0
MDL Number: MFCD17014743
Molecular Formula: C9H20BrN
Molecular Weight: 222.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 248
Formula:
C₉H₂₀BrN Melting Pt: 248 °C |
MDL Number:
MFCD17014743 CAS Number: 88840-42-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...