4-Methoxyphenyl-4-O-acetyl-3,6-di-O-benzyl-2-deoxy-2-phthalimido-β-D-glucopyranoside ≥98.0% (by HPLC)
Supplier: TCI America
M1834-1G
M1834-5G
TCM1834-1GEA
50.56
CAD
TCM1834-1G
TCM1834-5G
4-Methoxyphenyl-4-O-acetyl-3,6-di-O-benzyl-2-deoxy-2-phthalimido-β-D-glucopyranoside ≥98.0% (by HPLC)
4-Methoxyphenyl-4-O-acetyl-3,6-di-O-benzyl-2-deoxy-2-phthalimido-beta-D-glucopyranoside
CAS Number: 140615-77-6
Molecular Formula: C37H35NO9
Molecular Weight: 637.69
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 85 deg (C=0.8, CHCl3)
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Glycosylation, Bioscience, Glucosamine, Glycosides, p-Methoxyphenyl Glycosides, O-Substituted Sugars
Other Product Names / Synonyms: GlcNPhth[36Bn,4Ac]-beta-MP
Formula:
C₃₇H₃₅NO₉ |
CAS Number:
140615-77-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...